Masterflex® Large-Bore Slip Fittings
Larger bore for enhanced fluid flow
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These large-bore adapters are perfect for handling higher flow volumes in process while still using a slip-lock-style fitting system (similar to luer-style fittings). Adapters are manufactured in an ISO Class 8 (formerly Class 100,000) environment, are inspected to be free of contaminants, and are fully lot traceable. Packs of 25.
Polycarbonate:
Maximum temperature: 266°F (130°C).
Sterilization: Gamma irradiation, ETO.
Maximum temperature: 252°F (122°C).
Sterilization: Gamma irradiation, ETO.
Nylon:
Maximum temperature: 194°F (90°C).
Sterilization: Gamma irradiation, ETO, autoclave.
$41.50 - $50.50USD / Pkg of 25
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
COMPARE
3/16 Polycarbonate Large-bore adapter luer-style male x 3/16" hose barb, polycarbonate, pack of 25
Call For Price
COMPARE
1/4 Polycarbonate Large-bore adapter luer-style male x 1/4" hose barb, polycarbonate, pack of 25
$49.75USD / Pkg of 25
COMPARE
5/16 Polycarbonate Large-bore adapter luer-style male x 5/16" hose barb, polycarbonate, pack of 25
$49.75USD / Pkg of 25
COMPARE
3/16 Polycarbonate Luer Adapters, Female Luer to 3/16" ID, Large-Bore, PC, 25/Pk
$43.75USD / Pkg of 25
COMPARE
1/4 Polycarbonate Large-bore adapter luer-style female x 1/4" hose barb, polycarbonate, pack of 25
$45.75USD / Pkg of 25
COMPARE
Polycarbonate Connector cap for female large-bore, polycarbonate, pack of 25
$45.75USD / Pkg of 25
COMPARE
Polycarbonate Connector plug for male large-bore, polycarbonate, pack of 25
$46.75USD / Pkg of 25
COMPARE
3/16 Nylon Large-bore adapter luer-style male x 3/16" hose barb, nylon, pack of 25
$47.75USD / Pkg of 25
COMPARE
1/4 Nylon Large-bore adapter luer-style male x 1/4" hose barb, nylon, pack of 25
$47.75USD / Pkg of 25
COMPARE
5/16 Nylon Large-bore adapter luer-style male x 5/16" hose barb, pack of 25
$47.75USD / Pkg of 25
COMPARE
3/16 Nylon Large-bore adapter luer-style female x 3/16" hose barb, nylon, pack of 25
$41.50USD / Pkg of 25
COMPARE
1/4 Nylon Large-bore adapter luer-style female x 1/4" hose barb, nylon, pack of 25
$41.50USD / Pkg of 25
COMPARE
5/16 Nylon Large-bore adapter luer-style female x 5/16" hose barb, nylon, pack of 25
$41.50USD / Pkg of 25
COMPARE
Nylon Connector cap for female large-bore, nylon, pack of 25
$41.50USD / Pkg of 25
COMPARE
Nylon Connector plug for male large-bore, nylon, pack of 25
$42.25USD / Pkg of 25
COMPARE
Polycarbonate Male-male large-bore adapter, radiation-stable polycarbonate, pack of 25
$50.50USD / Pkg of 25
COMPARE
Polycarbonate Male-female large-bore adapter, radiation-stable polycarbonate, pack of 25
$50.50USD / Pkg of 25
Showing 1- 17 of 17