Masterflex® Large-Hose Peristaltic Pumps
Multiple designs—either dry pump cavity or lubricant filled—to best fit your system requirements
  • Complete isolation of pumped fluids
  • Self-priming and runs dry
  • Wide range of pump configurations and flows
  • No cups, packing, or seals for low maintenance
  • Will not break up delicate emulsions or create excessive frothing
The versatile VECTOR peristaltic pump series gives you pumping options you can’t get from standard positive displacement or centrifugal industrial pumps. The VECTOR pump is designed
to handle the high-viscosity fluids commonly found in industrial processing. Whether these fluids are pasty, pulpy or just plain thick, the VECTOR peristaltic pump can reliably start, stop, and continuously pump such fluids at a wide range of pressures and flows. These pumps are ideal for abrasive fluids, dyes, thick fluids, solids up 2.5 inches, shear-sensitive fluids, stringy material and corrosive fluids.
$6,090.00 - $47,600.00USD / Each
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connections Max Flow Rate (GPM) Max Pressure (PSI)
Availability Pricing
COMPARE
3" 316 SS ANSI 150-lb flange 23 100
$16,485.00USD / Each
COMPARE
3" 316 SS ANSI 150-lb flange 23 100
$17,055.00USD / Each
COMPARE
3" 316 SS ANSI 150-lb flange 23 100
$16,850.00USD / Each
COMPARE
3" 316 SS ANSI 150-lb flange 23 100
$17,055.00USD / Each
COMPARE
3" 316 SS ANSI 150-lb flange 39 100
$16,485.00USD / Each
COMPARE
3" 316 SS ANSI 150-lb flange 39 100
$16,850.00USD / Each
COMPARE
4" 316 SS ANSI 150-lb flange 57 100
$24,885.00USD / Each
COMPARE
4" 316 SS ANSI 150-lb flange 97 100
$24,625.00USD / Each
COMPARE
4" 316 SS ANSI 150-lb flange 97 100
$24,885.00USD / Each
COMPARE
4" 316 SS ANSI 150-lb flange 97 100
$25,140.00USD / Each
COMPARE
4" 316 SS ANSI 150-lb flange 97 100
$25,400.00USD / Each
COMPARE
6" 316 SS ANSI 150-lb flange 79 116
$45,645.00USD / Each
COMPARE
6" 316 SS ANSI 150-lb flange 79 116
$46,005.00USD / Each
COMPARE
6" 316 SS ANSI 150-lb flange 153 116
$46,985.00USD / Each
COMPARE
6" 316 SS ANSI 150-lb flange 153 116
$46,985.00USD / Each
COMPARE
6" 316 SS ANSI 150-lb flange 153 116
$46,985.00USD / Each
COMPARE
1.25" PVC hose barb 3.1 200
$6,090.00USD / Each
COMPARE
1.25" PVC hose barb 3.1 200
$6,233.00USD / Each
COMPARE
1.25" PVC hose barb 3.1 200
$6,275.00USD / Each
COMPARE
1.25" PVC hose barb 8.2 200
$6,244.00USD / Each
Showing 1 - 20 of 30