Masterflex® Liquid Flow Switches for High Inline Pressures
Ideal for high pressure applications
  • Withstands up to 1500 psi
  • Effortlessly install with 3/8" NPT(M) connections
The one-piece magnetic PPS composite piston makes these normally open switches ideal for high pressure applications. The single pole single throw (SPST) switches withstand up to 1500 psi at 70°F (21°C)! Use a 100 µm filter to protect your switch. Switches include 24 to 26" lead wires.
$169.70 - $396.40USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Housing Max Flow Rate (GPM) Process Connection (NPT(M))
Availability Pricing
COMPARE
Brass 0.5 3/8
$169.70USD / Each
COMPARE
Stainless Steel 0.25 3/8
$382.20USD / Each
COMPARE
Stainless Steel 2 3/8
$396.40USD / Each