This product is now discontinued, View Related Products
Masterflex® Liquid Flow Switches for Plastic Piping without Threads
Connects quickly to plastic piping
  • Consructed with PVC housings
Use these normally open switches with plastic pipes without threads. Switches are available in gray or clear PVC housing. Use clear switches where visual flow confirmation is needed. Use a 150 µm filter to protect your switch. Switches include 24" lead wires.
$126.20USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Pipe Size Wetted Materials
Availability Pricing
COMPARE
0.5 1/2" NPT PVC, Buna N, and epoxy
$126.20USD / Each
COMPARE
0.5 3/4" IPS PVC, Buna N, and epoxy
Call For Price
COMPARE
2 1" IPS PVC, Buna N, and epoxy
Call For Price
COMPARE
0.5 1/2" NPT PVC, Buna N, and epoxy
Call For Price
COMPARE
0.5 3/4" IPS PVC, Buna N, and epoxy
Call For Price
COMPARE
2 1" IPS PVC, Buna N, and epoxy