Masterflex® Liquid Flow Switches
Install switch at pipe elbow effortlessly
  • Durable switch with 225°F (107°C) max temperature and 250 psi at 70°F max pressure
  • Chemically resistant — wetted materials Ryton® PPS, Viton®, 316 SS, and epoxy
  • Effortlessly install with 9/16-18 UNF connections
No need to cut a straight pipe to install your flow switch. These SPDT switches with 90° ports easily fit where a pipe elbow would be installed. Use a 50 µm filter to protect your switch. Switches include 24" (61.0 cm) L leads.
$136.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Wetted Materials Description
Availability Pricing
COMPARE
0.1 Ryton® PPS, Viton®, 316 SS, and epoxy Liquid flow switch with 90° port; SPDT, 0.1 GPM
$136.00USD / Each
COMPARE
Ryton® PPS, Viton®, 316 SS, and epoxy Liquid flow switch with 90° port; SPDT, 0.25 GPM
$136.00USD / Each