Masterflex® Low-Flow Direct-Lift Proportioning Solenoid Valves
Resist pressures up to 500 psi!
  • Use for continuous control of liquid or gas flow
  • Valves withstand pressures to 500 psi
Apply a 0 to 30 VDC input signal—these normally-closed valves open to give you flow rates that are proportional to the input voltage. Stainless steel valve construction withstands pressures to 500 psi. Maximum fluid temperature is 122°>F (50°>C). Maximum ambient temperature with intermittent use is 130°>F (54°>C). Actuation time is 500 milliseconds.
Wetted parts are 316 and 416 stainless steel and VITON®. Ports are 1/4" compression fittings. Valves include 6" lead wires.
Optional Control Module allows valves to accept 0 to 5 VDC or 4 to 20 mA input signal—ideal for use with transmitters or controllers. Control module has a DB9 female connector (cord sold separately) for input/output signals and requires a 12 to 30 VDC power supply.
$340.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate For Air (mL/min) Max Flow Rate For Water (mL/min) Orifice (in)
Availability Pricing
COMPARE
3500 125 0.02
$340.00USD / Each
COMPARE
13000 400 0.04
$340.00USD / Each
COMPARE
21500 700 0.055
$340.00USD / Each
COMPARE
25000 850 0.063
$340.00USD / Each
COMPARE
100000 2873 0.125
$340.00USD / Each
Showing 1- 5 of 5
Need Help with this product?
Call Us At1-847-549-7600
Live chat