Masterflex® Low-Flow Peristaltic Pumps
Variable-flow reversible peristaltic pump
  • Flow ranges from 0.03 to 4 mL/min
  • Run dry without damage to pump
These cost-effective peristaltic pumps provide variable flow and exceptional life performance in a compact package. Select from three different motor speeds, and three different tubing sizes to achieve flow rates from 0.03 to 4 mL/min.
Order power supply separately. Wall mount power supply, 115 VAC/12 VDC with US standard plug sold separately as part number 73160-50. Wall mount power supply, 230 VAC/12 VDC with EU plug sold separately as part number 73160-52.
$255.90 - $368.20USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Description
Availability Pricing
COMPARE
1 0.03 Low Flow Peristaltic Pump, Variable Speed, 1.5 - 5 RPM, 12 VDC, Manual Control
$261.10USD / Each
COMPARE
1 0.03 Peristaltic Pump, Reversible, Remote; 0.03-1 mL/min, 12 VDC
$365.10USD / Each
COMPARE
1.9 0.06 Low Flow Peristaltic Pump, Variable Speed, 3.3 - 10 RPM, 12 VDC, Manual Control
$262.10USD / Each
COMPARE
1.9 0.06 Peristaltic Pump, Reversible, Remote; 0.06-1.9 mL/min, 12 VDC
$340.10USD / Each
COMPARE
4 0.14 Low Flow Peristaltic Pump, Variable Speed, 8 - 22 RPM, 12 VDC, Manual Control
$255.90USD / Each
COMPARE
4 0.14 Peristaltic Pump, Reversible, Remote; 0.14-4 mL/min, 12 VDC
$368.20USD / Each