Masterflex® Low Pressure Drop Gas Mass Flowmeter for Gases
Achieve high accuracy and low pressure drop!
  • Full-scale pressure drops as low as 0.1 psid to 0.69 psid
  • Measure 30 standard gases with a 200:1 turndown ratio
  • Set up and control from an easy-to-use keypad
Using a newer laminar differential pressure flow technology, these mass flow meters and controllers have a much lower pressure drop across the instrument. Meters and controllers require no warm-up time, and offer a 10 ms response time for meters and 100 ms for controllers. Typical applications include air sampling, natural gas metering and calibration, and gas analysis and totalizing. Select from 130 different gases in real time with the gas and gas mix calibrations already programmed into the meter or controller. An integrated keypad around the display is all that is required to program the unit for service. The 0 to 5 VDC output permits transmission of the flow value to a remote display, recorder, or controller regulating a valve or pump.
Battery-Powered Meters allow you to take the meter from site to site for flow rate establishment and on-site calibration. They have a monochrome display and rechargeable integrated battery packs.
Voltage-Powered Meters and Controllers are designed for single-area use and feature a dynamic, color thin-film transistor (TFT) display with backlight. The TFT display simultaneously shows flow rate, line pressure, fluid temperature, and (for controllers) the set point. Easily see when the unit is at max range or out of range—each numeric reading of the four parameters changes to yellow at 100% of range or red at 128% of range. Both power and input/output signals are transmitted through a single multipin connector.
$1,925.00 - $4,944.00USD / Each
47 variations of this product are available.
See All Product Options Below
47 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max Flow Rate (LPM) Process Connection
Availability Pricing
COMPARE
7-line x 4-digit LCD; 1.25"H flow display 0.0005 10-32 UNF
$2,559.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display 0.001 10-32 UNF
$2,559.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display 1 1/8" NPT(F)
$1,941.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display 10 1/4" NPT(F)
$2,013.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display 20 1/4" NPT(F)
$2,023.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display 100 3/4" NPT(F)
$3,023.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display 250 3/4" NPT(F)
$3,207.00USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display 500 3/4" NPT(F)
$4,365.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 0.0005 10-32 UNF
$2,630.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 0.001 10-32 UNF
$2,652.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 0.005 10-32 UNF
$2,399.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 0.5 1/8" NPT(F)
$1,940.00USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 5 1/4" NPT(F)
$2,023.00USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 20 1/4" NPT(F)
$2,091.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 40 1/2" NPT(F)
$2,409.50USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 250 3/4" NPT(F)
$3,306.00USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 0.05 1/8" NPT(F)
$2,541.00USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 0.2 1/8" NPT(F)
$2,405.00USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 1 1/8" NPT(F)
$2,418.00USD / Each
COMPARE
7-line x 4-digit LCD; 1.25"H flow display, bright TFT 50 3/4" NPT(F)
$3,750.00USD / Each
Showing 1 - 20 of 47