Masterflex® Mechanically Coupled Self-Priming Centrifugal Pumps
Chemical-resistance pump heads provide dependable performance and continuous service
  • Mechanically close-coupled design saves space and simplifies installation and maintenance
  • Back pullout construction — assembly and overhaul of impeller and seal without distorting suction and discharge connections
  • Top centerline discharge and foot support under casing ensure self-venting and reduce misalignment from pipe loads
The mechanically coupled centrifugal pumps, constructed of 304 stainless steel, are ideal for food and pharmaceutical processing plants, as well as water and wastewater treatment centers. They feature high-quality mechanical shaft seals and O-rings. Also available are optional materials for high-temperature and chemical-duty operations. Contact an Application Specialist for more information.
$1,054.00 - $1,191.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Power (VAC)
Availability Pricing
COMPARE
19 120 115 / 230
$1,054.00USD / Each
COMPARE
18 180 115 / 230
$1,191.00USD / Each
COMPARE
18 180 230 / 460
$1,121.00USD / Each