Masterflex® Metal Metering Valves
Choose from different metal materials based on your application
  • Brass and 316 Stainless Steel material options available
Brass valves feature Buna N O-rings; stainless steel valves have Viton® O-rings.
$62.00 - $155.00USD / Each
21 variations of this product are available.
See All Product Options Below
21 PRODUCT OPTIONS
Compareitem specifications
Description Max Flow Rate For Air (mL/min) Max Flow Rate For Water (mL/min)
Availability Pricing
COMPARE
High-Resolution Brass Metering Valve, straight flow, 6 mL/min 200 6
$94.00USD / Each
COMPARE
High-Resolution Brass Metering Valve, straight flow, 12 mL/min 400 12
$94.00USD / Each
COMPARE
High-Resolution Brass Metering Valve, straight flow, 30 mL/min 1000 30
$94.00USD / Each
COMPARE
High-Resolution Brass Metering Valve, straight flow, 200 mL/min 6200 200
$94.00USD / Each
COMPARE
High-Resolution Brass Metering Valve, straight flow, 650 mL/min 21500 650
$94.00USD / Each
COMPARE
High-Resolution 316 SS Metering Valve, straight flow, 6 mL/min 200 6
$155.00USD / Each
COMPARE
High-Resolution 316 SS Metering Valve, straight flow, 12 mL/min 400 12
$155.00USD / Each
COMPARE
High-Resolution 316 SS Metering Valve, straight flow, 30 mL/min 1000 30
$155.00USD / Each
COMPARE
High-Resolution 316 SS Metering Valve, straight flow, 70 mL/min 2500 70
$155.00USD / Each
COMPARE
High-Resolution 316 SS Metering Valve, straight flow, 200 mL/min 6200 200
$155.00USD / Each
COMPARE
High-Resolution 316 SS Metering Valve, straight flow, 650 mL/min 21500 650
$155.00USD / Each
COMPARE
Brass Metering Valve, straight flow, 350 mL/min 5000 350
$62.00USD / Each
COMPARE
316 SS Metering Valve, straight flow, 350 mL/min 5000 350
$101.00USD / Each
COMPARE
Brass Metering Valve, 90 deg flow, 350 mL/min 5000 350
$64.00USD / Each
COMPARE
316 SS Metering Valve, 90 deg flow, 350 mL/min 5000 350
$103.00USD / Each
COMPARE
316 SS Metering Valve, straight flow, 1200 mL/min 20000 1200
$101.00USD / Each
COMPARE
316 SS Metering Valve, 90 deg flow, 1200 mL/min 220000 1200
$103.00USD / Each
COMPARE
Brass Metering Valve, straight flow, 3500 mL/min 60000 3500
$62.00USD / Each
COMPARE
316 SS Metering Valve, straight flow, 3500 mL/min 60000 3500
$101.00USD / Each
COMPARE
316 SS Metering Valve, 90 deg flow, 3500 mL/min 60000 3500
$103.00USD / Each
Showing 1 - 20 of 21