Masterflex® Microscope Slides with Wells
Keep samples isolated
  • Screen-printed well design prevents bleed-through
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Contrasting ink makes orienting the microscope field of view over a sample easy
These printed microscope slides feature circular wells designed to hold samples in place, allowing multiple wells per slide, so you can increase throughput without adding to cleanup. Black PTFE-based epoxy makes locating wells with a microscope simple and quick, improving efficiency. Select from a range of well counts to suit any application.
$67.50 - $75.00USD / Pkg of 144
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Slide Type Description
Availability Pricing
COMPARE
Printed with Mask Printed Microscope Slide, one well; 144/pk
$71.00USD / Pkg of 144
COMPARE
Printed with Mask Printed Microscope Slide, two well; 144/pk
$71.00USD / Pkg of 144
COMPARE
Printed with Mask Printed Microscope Slide, three well; 144/pk
$71.00USD / Pkg of 144
COMPARE
Printed with Mask Printed Microscope Slide, four well; 144/pk
$68.50USD / Pkg of 144
COMPARE
Printed with Mask Printed Microscope Slide, six well; 144/pk
$67.50USD / Pkg of 144
COMPARE
Printed with Mask Printed Microscope Slide, eight well; 144/pk
$75.00USD / Pkg of 144
COMPARE
Printed with Mask Printed Microscope Slide, 48 well; 144/pk
$71.00USD / Pkg of 144
Showing 1- 7 of 7