Masterflex® Motorized Drum Pumps
Complete drum pump system specifically packaged for different applications
  • Flow rate 35 GMP for all models
  • Equipped with variable-speed control motors for added safety and control
  • Maintenance is reduced with no mechanical seals
  • Run dry capability
Pre-configured drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks, and small storage vessels. Based upon the application, polypropylene (PP), CPVC, PVDF, and 316 stainless steel (SS) materials provide excellent chemical resistance against corrosive liquids.
The Measurement Pump Packages (Models 70614-06, -08 and -09) include a digital flowmeter and totalizer. The dispensing hose for all the pumps is a 6-ft length of PVC tubing. The storage bracket for all packages is made from 16 gauge coated steel.
$1,507.00 - $5,204.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Tube Length, Inlet (in) Description
Availability Pricing
COMPARE
39 Motorized Drum Pump Package, 35 GPM, 39"L CPVC Tube; 115V
$1,585.00USD / Each
COMPARE
39 Motorized Drum Pump Package, 35 GPM, 39"L CPVC Tube; 230V
$1,617.00USD / Each
COMPARE
39 Motorized Drum Pump Package, 35 GPM, 39"L PP Tube; 115V
$1,507.00USD / Each
COMPARE
39 Motorized Drum Pump Package, 35 GPM, 39"L PP Tube; 230V
$1,570.00USD / Each
COMPARE
39 Motorized Drum Pump Package, 35 GPM, 39"L PVDF Tube; 115V
$2,380.00USD / Each
COMPARE
39 Motorized Drum Pump Package, 35 GPM, 39"L PVDF Tube; 230V
$2,396.00USD / Each
COMPARE
39 Drum Pump System w/Flow Monitor, 35 GPM, 39"L PP Tube; 115V
$3,559.00USD / Each
COMPARE
39 Drum Pump System w/Flow Monitor, 35 GPM, 39"L PVDF Tube; 115V
$5,115.00USD / Each
COMPARE
39 Drum Pump System w/Flow Monitor, 35 GPM, 39"L PVDF Tube; 230V
$5,204.00USD / Each
COMPARE
39 Motorized Drum Pump System, 35 GPM, 39"L SS Tube; 115V
$2,006.00USD / Each
Showing 1- 10 of 10