Masterflex® Multi-Syringe Pumps, Multi-Step
Ideal for low-flow, low-pressure applications where precision is needed
  • Flow ranges from 0.05 uL/min up to 70.56 mL/min
  • Built-in program lets you enter up to eight commands to perform a variety of functions—you can even repeat commands up to ten times; all settings are stored in nonvolatile memory
  • Backlit alphanumeric LCD ste
Program flow rate hold, pause, and change of direction; each command can be set to run for up to 12 hours. You can even program infusion/withdrawal pumps to sequence operations: either withdrawal followed by infusion or infusion followed by withdrawal. Ch
$5,009.00 - $6,711.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Syringes Power (VAC) Description
Availability Pricing
COMPARE
2 115 Infusion Pump, Dual-Syringe, Programmable; 115 VAC
$5,009.00USD / Each
COMPARE
2 230 Infusion Pump, Dual-Syringe, Programmable; 230 VAC
$5,247.00USD / Each
COMPARE
10 115 Infusion Pump, Ten-Syringe Capacity; 115 VAC 50/60 Hz
$5,765.00USD / Each
COMPARE
10 230 Infusion Pump, Ten-Syringe Capacity; 230 VAC 50/60 Hz
$5,962.00USD / Each
COMPARE
2 115 Infusion/Withdrawal Pump, Dual-Syringe, Programmable; 115 VAC
$5,758.00USD / Each
COMPARE
10 230 Infusion/Withdrawal Pump, Ten-Syringe, Programmable; 230 VAC
$6,711.00USD / Each