Masterflex® Multi-Syringe Pumps
Economical & packed with features
  • Flow rates up to 147 mL/min per syringe
  • Choose pumps which accept two or up to ten syringes—ideal for pumping exact proportions
  • RS-232 interface lets you easily control pumps from your computer
  • Backlit alphanumeric LCD steps you through setup
Simple keypad programming will have you up and running quickly. Program infusion/withdrawal pumps to sequence operations: either withdrawal followed by infusion or infusion followed by withdrawal.
Pumps feature two TTL inputs and two TTL outputs. The TTL
$3,444.50 - $4,738.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate Per Syringe (mL/min) Number of Syringes Description
Availability Pricing
COMPARE
147 2 Dual-syringe infusion pump, 230 VAC
$3,444.50USD / Each
COMPARE
147 2 Dual-syringe infusion/withdrawal pump, 230 VAC
$4,725.50USD / Each
COMPARE
147 10 Ten-syringe infusion pump, 230 VAC
$4,738.50USD / Each