Masterflex® Multi-Turn PTFE Needle Valves
Easily meter corrosive fluids or gases
Maximum pressure is 75 psi; maximum temperature is 200°>F (93°>C). Wetted parts are body and O-ring made of PTFE and CTFE valve spindle. Choose aluminum (Al) or 316 stainless steel (SS) body. Orifice is 0.125".
$74.00 - $254.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Max Flow Rate For Water (mL/min) Wetted Materials
Availability Pricing
COMPARE
1/8" NPT(F) 2800 Aluminum, PTFE, CTFE
$74.00USD / Each
COMPARE
3/8" tubing connector 130 Aluminum, PTFE, CTFE
$79.00USD / Each
COMPARE
1/4" compression 130 Aluminum, PTFE, CTFE
$224.00USD / Each
COMPARE
1/4" compression 2800 Aluminum, PTFE, CTFE
$224.00USD / Each
COMPARE
1/8" NPT(F) 130 Aluminum, PTFE, CTFE
$74.00USD / Each
COMPARE
3/8" tubing connector 2800 Aluminum, PTFE, CTFE
$79.00USD / Each
COMPARE
1/8" NPT(F) 130 Stainless Steel, PTFE, CTFE
$99.00USD / Each
COMPARE
1/8" NPT(F) 2800 Stainless Steel, PTFE, CTFE
$99.00USD / Each
COMPARE
3/8" tubing connector 130 Stainless Steel, PTFE, CTFE
$109.00USD / Each
COMPARE
3/8" tubing connector 2800 Stainless Steel, PTFE, CTFE
$109.00USD / Each
COMPARE
1/4" compression 130 Stainless Steel, PTFE, CTFE
$254.00USD / Each
COMPARE
1/4" compression 2800 Stainless Steel, PTFE, CTFE
$254.00USD / Each
Showing 1- 12 of 12
Need Help with this product?
Call Us At1-847-549-7600
Live chat