Masterflex® Normally Open Flow Switch for Low Pressure-Drop Liquids
Minimizes flow constriction
  • Provides low pressure drop
These SPST switches feature a unique, dual-diameter, internal bore and piston configuration to minimize flow constriction and provide low pressure drop. Use a 150 um filter to protect your switch.
$129.70 - $186.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Power Wetted Materials
Availability Pricing
COMPARE
0.5 20 VA PP, Ryton® PPS, Viton®, and 316 SS
$129.70USD / Each
COMPARE
1 20 VA PP, Ryton® PPS, Viton®, and 316 SS
$186.00USD / Each
COMPARE
2 20 VA PP, Ryton® PPS, Viton®, and 316 SS
$181.50USD / Each
COMPARE
3 20 VA PP, Ryton® PPS, Viton®, and 316 SS
$129.70USD / Each