Masterflex® Normally Open Low-Flow Compact Switches
Provides reliable operation in clean air and water applications
  • Constructed of Noryl®, 316 stainless steel, and epoxy
These normally open SPST switches are specifically designed for reliable operation in clean air and water applications. Select from gas or liquid flow switches. Use a 50-µm filter to protect your switch. Switches include 18" (45.7 cm) L leads.
$132.50 - $186.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Media Type Wetted Materials Description
Availability Pricing
COMPARE
Noryl®, 316 stainless steel, and epoxy Compact low-flow switch for gases; SPST normally open, 25 cfh/60 cfh
$186.00USD / Each
COMPARE
Noryl®, 316 stainless steel, and epoxy Compact low-flow switch for liquids; SPST normally open, 0.05 GPM
$186.00USD / Each
COMPARE
Noryl®, 316 stainless steel, and epoxy Compact low-flow switch for gases; SPST normally open, 1 cfm/2.6 cfm
$186.00USD / Each
COMPARE
Noryl®, 316 stainless steel, and epoxy Compact low-flow switch for liquids; SPST normally open, 0.25 GPM
$186.00USD / Each
COMPARE
Noryl®, 316 stainless steel, and epoxy Compact low-flow switch for gases; SPST normally open, 2.5 cfm/5.6 cfm
$132.50USD / Each
COMPARE
Noryl®, 316 stainless steel, and epoxy Compact low-flow switch for liquids; SPST normally open, 0.5 GPM
$186.00USD / Each
COMPARE
Noryl®, 316 stainless steel, and epoxy Compact low-flow switch for gases; SPST normally open, 5 cfm/12 cfm
$133.50USD / Each
Showing 1- 7 of 7