Masterflex® Open-Head Sensor Interlock Retrofit System Accessories
Make the most of your Masterflex® Open-Head Sensor with these accessories and replacement parts
  • Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
  • Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
  • Controller accepts input signals from up to two pump heads
  • Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
  • Controller is NEMA 4X / IP66 rated for washdown
The retrofit system is compatible with all I/P® pumps and with L/S® pumps that accept the Easy-Load® II pump head.
Use only with I/P® Easy-Load® pump head 77602-30 and with L/S® Easy-Load® II pump heads 77200-30 and 77200-32.
$31.00 - $93.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Replacement Masterflex Open-Head Sensor Shorting Plug for Sensor-Equipped Drives
$33.00USD / Each
COMPARE
Power Cable for Masterflex Open-Head Sensor Interface; 115 V, 6 ft (1.8 m) L
$31.00USD / Each
COMPARE
Extension Cable for Masterflex Open-Head Sensor Interface; 6 ft (1.8 m) L
$93.50USD / Each