Masterflex® Open Impeller Centrifugal Pumps
Suitable for suspended solids
  • Close-coupled design offers smallest possible footprint to save space
  • Back-pullout construction allows for servicing of pump without disturbing inlet and outlet piping
  • Top centerline discharge ensures self-venting of entrapped gases
Typical applications include food process, cooling systems, beverage processing and pharmaceutical services. The open impeller design means the pump can easily handle suspended solids up to a 3/4" spherical shape. Pumps are non-overloading and rated for continuous duty throughout entire operating range for reliable performance and long life.
$1,226.50 - $1,389.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Max Motor RPM (rpm)
Availability Pricing
COMPARE
155 33 3450
$1,226.50USD / Each
COMPARE
175 46 3450
$1,310.00USD / Each
COMPARE
230 53 3450
$1,389.00USD / Each