Masterflex Paddle Wheel Flowmeters for Monitoring and Control
Meters provide visual and remote flow indication for numerous applications
  • Choose from standard or digital models with display and keypad
  • Manufactured from solid PP or PVDF; choose PVDF for better chemical resistance
  • Jewel bearings allow for very low minimum flow rates
  • Only one moving part provides extended life
Designed for water-based liquids, these IP65-rated flowmeters provide flow measurement to 35 GPM (132.5 LPM) and models with the RTD option measure temperature to 140°F (60°C). The RTD is platinum encased in 316 stainless steel and has a 4-wire 12" (30.5 cm) cable for output. Flow indication can be seen via acrylic cover. Mount units horizontally or vertically.
$368.00 - $1,260.00USD / Each
32 variations of this product are available.
See All Product Options Below
32 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Pipe Size Wetted Materials
Availability Pricing
COMPARE
5 3/8" NPT (F) PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
$368.00USD / Each
COMPARE
10 1/2" NPT (F) PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
$368.00USD / Each
COMPARE
15 3/4" NPT (F) PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
$368.00USD / Each
COMPARE
5 3/8" NPT (F) PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
$513.00USD / Each
COMPARE
10 1/2" NPT (F) PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
$513.00USD / Each
COMPARE
5 3/8" NPT (F) PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
$727.00USD / Each
COMPARE
17 3/4" NPT (F) PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
$727.00USD / Each
COMPARE
5 3/8" NPT (F) PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
$880.00USD / Each
COMPARE
17 3/4" NPT (F) PP, Acrylic, PVDF, nickel tungsten carbide, sapphire, EPDM, and 316 SS
$880.00USD / Each
COMPARE
5 3/8" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, and 316 SS
$745.00USD / Each
COMPARE
10 1/2" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
$745.00USD / Each
COMPARE
35 1" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
$745.00USD / Each
COMPARE
5 3/8" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
$891.00USD / Each
COMPARE
10 1/2" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
$891.00USD / Each
COMPARE
17 3/4" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
$891.00USD / Each
COMPARE
10 1/2" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
$1,110.00USD / Each
COMPARE
17 3/4" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
$1,110.00USD / Each
COMPARE
35 1" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
$1,110.00USD / Each
COMPARE
10 1/2" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
$1,260.00USD / Each
COMPARE
17 3/4" NPT (F) PVDF, zirconia ceramic, saphhire, PTFE, 316 SS
$1,260.00USD / Each
Showing 1 - 20 of 32
Need Help with this product?
Call Us At1-847-549-7600
Live chat