Masterflex® Paddle-Wheel Micro-Flo Rate and Total Flowmeters
Economical low-flow ranges
  • Paddle wheel technology is accurate, affordable, and relatively easy to install and maintain
  • Ideally suited to open channels and partially filled pipes containing water
  • Displays flow rate and total
  • Preset factory calibrations or custom field-calibrated
These economical flowmeters are capable of very low flow rates. The display is programmable for preselected factory settings or can be field calibrated for high accuracy in a specific application. Displays units in milliliters, ounces, liters, or gallons; displays time in minutes, hours, or days. Display can be programmed for up to four decimal places. All units are supplied with a 115 VAC/DC power supply.
$514.00 - $528.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Process Connection
Availability Pricing
COMPARE
300 30 1/4" NPT(F)
$520.00USD / Each
COMPARE
1000 100 1/4" NPT(F)
$514.00USD / Each
COMPARE
2000 200 1/4" NPT(F)
$524.00USD / Each
COMPARE
2000 300 1/4" NPT(F)
$514.00USD / Each
COMPARE
5000 500 1/4" NPT(F)
$524.00USD / Each
COMPARE
7000 700 1/4" NPT(F)
$514.00USD / Each
COMPARE
300 30 1/4" ID x 3/8" OD tubing connection
$528.00USD / Each
COMPARE
1000 100 1/4" ID x 3/8" OD tubing connection
$528.00USD / Each
COMPARE
2000 200 1/4" ID x 3/8" OD tubing connection
$514.00USD / Each
COMPARE
3000 300 1/4" ID x 3/8" OD tubing connection
$514.00USD / Each
COMPARE
5000 500 1/4" ID x 3/8" OD tubing connection
$528.00USD / Each
COMPARE
3000 70 1/4" ID x 3/8" OD tubing connection
$528.00USD / Each
Showing 1- 12 of 12
Need Help with this product?
Call Us At1-847-549-7600
Live chat