Masterflex® Panel-Mount Direct-Reading Variable-Area Flowmeters for Air, 150-mm Scale
No more eye strain—front shield magnifies scale 16 percent for easier readings
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
$176.40 - $320.80USD / Each
46 variations of this product are available.
See All Product Options Below
46 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Max Flow Rate (mL/min) Description
Availability Pricing
COMPARE
Aluminum 75 150-mm Direct Reading Flowmeter, Al/Cb Float; Air, 75 mL/min
$183.50USD / Each
COMPARE
Aluminum 200 150-mm Direct Reading Flowmeter, Al/SS Float; Air, 200 mL/min
$183.50USD / Each
COMPARE
Aluminum 5000 150-mm Direct Reading Flowmeter, Al/Saph Float; Air, 5 LPM
$183.50USD / Each
COMPARE
Aluminum 10000 150-mm Direct Reading Flowmeter, Al/Gl Float; Air, 10 LPM
$183.50USD / Each
COMPARE
Aluminum 10854 150-mm Direct Reading Flowmeter, Al/Cb Float; Air, 23 scfh
$183.50USD / Each
COMPARE
Chrome-plated brass 200 150-mm Direct Reading Flowmeter, Br/SS Float; Air, 200 mL/min
$194.70USD / Each
COMPARE
Chrome-plated brass 300 150-mm Direct Reading Flowmeter, Br/Gl Float; Air, 300 mL/min
$194.70USD / Each
COMPARE
Chrome-plated brass 1179 150-mm Direct Reading Flowmeter, Br/Cb Float; Air, 2.5 scfh
$194.70USD / Each
COMPARE
Chrome-plated brass 3893 150-mm Direct Reading Flowmeter, Br/Gl Float; Air, 8.25 scfh
$194.70USD / Each
COMPARE
Chrome-plated brass 23597 150-mm Direct Reading Flowmeter, Br/Gl Float; Air, 50 scfh
$201.60USD / Each
COMPARE
316 SS 2500 150-mm Direct Reading Flowmeter, SS/Gl Float; Air, 2.5 LPM
$308.70USD / Each
COMPARE
316 SS 10000 150-mm Direct Reading Flowmeter, SS/Gl Float; Air, 10 LPM
$308.70USD / Each
COMPARE
Aluminum 25 150-mm Direct Reading Flowmeter, Al/Saph Float; Air, 25 mL/min
$183.50USD / Each
COMPARE
Aluminum 500 150-mm Direct Reading Flowmeter, Al/Saph Float; Air, 500 mL/min
$183.50USD / Each
COMPARE
Aluminum 800 150-mm Direct Reading Flowmeter, Al/Gl Float; Air, 800 mL/min
$183.50USD / Each
COMPARE
Aluminum 60000 150-mm Direct Reading Flowmeter, Al/Cb Float; Air, 60 LPM
$196.60USD / Each
COMPARE
Chrome-plated brass 500 150-mm Direct Reading Flowmeter, Br/Saph Float; Air, 500 mL/min
$194.70USD / Each
COMPARE
Chrome-plated brass 23000 150-mm Direct Reading Flowmeter, Br/Gl Float; Air, 23 LPM
$205.70USD / Each
COMPARE
316 SS 1.25 150-mm Direct Reading Flowmeter, SS/Cb Float; Air, 1.25 mL/min
$308.70USD / Each
COMPARE
316 SS 42000 150-mm Direct Reading Flowmeter, SS/SS Float; Air, 42 LPM
$320.80USD / Each
Showing 1 - 20 of 46