Masterflex® Panel-Mount Direct-Reading Variable-Area Flowmeters for Air and Water, 65-mm Scale
No more eye strain - front shield magnifies scale 16 percent for easier readings
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
$113.30 - $255.60USD / Each
38 variations of this product are available.
See All Product Options Below
38 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Valve Description
Availability Pricing
COMPARE
Aluminum No 65-mm Direct Reading Flowmeter, Aluminum/SS Float; For Air, 50 mL/min
$113.30USD / Each
COMPARE
Aluminum No 65-mm Flowmeter, Aluminum/Carboloy Float; For Air, 250 mL/min
$113.30USD / Each
COMPARE
Aluminum No 65-mm Direct Reading Flowmeter, Aluminum/Glass Float; For Air, 1 LPM
$113.30USD / Each
COMPARE
Aluminum No 65-mm Direct Reading Flowmeter, Aluminum/Glass Float; For Air, 7 mL/min
$113.30USD / Each
COMPARE
Aluminum No 65-mm Direct Reading Flowmeter, Aluminum/Ss Float; For Air, 10 LPM
$113.30USD / Each
COMPARE
Aluminum No 65-mm Flowmeter, Aluminum/Glass Float; For Water, 500 mL/min
$124.30USD / Each
COMPARE
Chrome-plated brass No 65-mm Direct Reading Flowmeter, Brass/Ss Float; For Air, 50 mL/min
$145.50USD / Each
COMPARE
Chrome-plated brass No 65-mm Flowmeter, Brass/Carboloy Float; For Air, 500 mL/min
$145.50USD / Each
COMPARE
Chrome-plated brass No 65-mm Direct Reading Flowmeter, Brass/Glass Float; For Air, 1 LPM
$145.50USD / Each
COMPARE
Chrome-plated brass No 65-mm Flowmeter, Brass/Ss Float; For Air, Without Valve; 2 LPM
$145.50USD / Each
COMPARE
Chrome-plated brass No 65-mm Direct Reading Flowmeter, Brass/Glass Float; For Air, 5 LPM
$145.50USD / Each
COMPARE
Chrome-plated brass No 65-mm Direct Reading Flowmeter, Brass/Ss Float; For Air, 6 mL/min
$145.50USD / Each
COMPARE
Chrome-plated brass No 65-mm Direct Reading Flowmeter, Brass/Ss Float; For Air, 115 mL/min
$145.50USD / Each
COMPARE
316 SS No 65-mm Direct Reading Flowmeter, Ss/Glass Float; For Air, 100 mL/min
$250.80USD / Each
COMPARE
316 SS No 65-mm Direct Reading Flowmeter, Ss/Carboloy Float; For Air, 250 mL/min
$250.80USD / Each
COMPARE
316 SS No 65-mm Direct Reading Flowmeter, Ss/Carboloy Float; For Air, 500 mL/min 32012-07
$250.80USD / Each
COMPARE
316 SS No 65-mm Flowmeter, Ss/Ss Float; For Air, Without Valve; 2 LPM
$250.80USD / Each
COMPARE
316 SS No 65-mm Flowmeter, Ss/Ss Float; For Air, Without Valve; 10 LPM
$250.80USD / Each
COMPARE
316 SS No 65-mm Direct Reading Flowmeter, Ss/Glass Float; For Air, 6 scfh
$250.80USD / Each
COMPARE
316 SS No 65-mm Direct Reading Flowmeter w/o Valve, SS, 0.5 mL/min H2O
$250.80USD / Each
Showing 1 - 20 of 38