Masterflex® Panel-Mount Direct-Reading Variable-Area Flowmeters for Routine Gases, Aluminum, 150-mm Scale
Ideal for gas applications where metal components must be minimized
  • Transparent front safety shield enhances reading resolution by 16%
  • Rugged design features case-enclosed flowtube
  • Tubes include a fused ceramic scale as a precise, permanent measuring guide
These meters eliminate the need for correlation charts with direct-reading scales for individual routine gases. The inert PTFE and glass wetted parts combine with the rigid anodized aluminum frame to offer a unique solution for ultra-pure gas applications. Flowtubes are replaceable.
$337.80 - $441.00USD / Each
27 variations of this product are available.
See All Product Options Below
27 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Gas Type Max Flow Rate (LPM) Valve
Availability Pricing
COMPARE
Argon 0.033 Yes
$427.90USD / Each
COMPARE
Argon 15 Yes
$427.90USD / Each
COMPARE
Helium 0.1 Yes
$427.90USD / Each
COMPARE
Helium 0.5 Yes
$427.90USD / Each
COMPARE
Helium 1.5 Yes
$427.90USD / Each
COMPARE
Helium 5 Yes
$427.90USD / Each
COMPARE
Helium 40 Yes
$427.90USD / Each
COMPARE
Nitrogen 0.3 Yes
$427.90USD / Each
COMPARE
Nitrogen 0.5 Yes
$427.90USD / Each
COMPARE
Nitrogen 2 Yes
$427.90USD / Each
COMPARE
Nitrogen 0.76 Yes
$441.00USD / Each
COMPARE
Oxygen 0.25 Yes
$427.90USD / Each
COMPARE
Oxygen 5 Yes
$427.90USD / Each
COMPARE
Oxygen 16.5 Yes
$427.90USD / Each
COMPARE
Oxygen 58 Yes
$441.00USD / Each
COMPARE
Argon 0.033 No
$337.80USD / Each
COMPARE
Nitrogen 0.1 No
$337.80USD / Each
COMPARE
Nitrogen 0.2 No
$337.80USD / Each
COMPARE
Oxygen 0.25 No
$337.80USD / Each
COMPARE
Oxygen 0.4 No
$337.80USD / Each
Showing 1 - 20 of 27