Masterflex® Panel-Mount Direct-Reading Variable-Area Flowmeters for Routine Gases, Aluminum, 65-mm Scale
Ideal for gas applications where metal components must be minimized
  • Transparent front safety shield enhances reading resolution by 16%
  • Rugged design features case-enclosed flowtube
  • Tubes include a fused ceramic scale as a precise, permanent measuring guide
These meters eliminate the need for correlation charts with direct-reading scales for individual routine gases. The inert PTFE and glass wetted parts combine with the rigid anodized aluminum frame to offer a unique solution for ultra-pure gas applications. Flowtubes are replaceable.
$249.60 - $380.70USD / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (mL/min) Min Flow Rate (mL/min) Wetted Materials
Availability Pricing
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$344.80USD / Each
COMPARE
$380.70USD / Each
COMPARE
$249.60USD / Each
COMPARE
$249.60USD / Each
COMPARE
$249.60USD / Each
COMPARE
$249.60USD / Each
COMPARE
$249.60USD / Each
COMPARE
$249.60USD / Each
Showing 1- 20 of 20