Masterflex® Panel-Mount Direct-Reading Variable-Area Flowmeters for Water, 150-mm Scale
No more eye strain—front shield magnifies scale 16 percent for easier readings
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
$183.50 - $346.80USD / Each
38 variations of this product are available.
See All Product Options Below
38 PRODUCT OPTIONS
Compareitem specifications
Material Fitting Max Flow Rate (mL/min) Valve
Availability Pricing
COMPARE
Aluminum 10 No
$183.50USD / Each
COMPARE
Aluminum 100 No
$183.50USD / Each
COMPARE
Aluminum 500 No
$183.50USD / Each
COMPARE
Chrome-plated brass 10 No
$194.70USD / Each
COMPARE
Chrome-plated brass 20 No
$194.70USD / Each
COMPARE
Chrome-plated brass 500 No
$194.70USD / Each
COMPARE
316 SS 100 No
$308.70USD / Each
COMPARE
316 SS 500 No
$308.70USD / Each
COMPARE
Aluminum 10 Yes
$194.50USD / Each
COMPARE
Aluminum 20 Yes
$200.70USD / Each
COMPARE
Aluminum 500 Yes
$200.70USD / Each
COMPARE
Chrome-plated brass 10 Yes
$224.70USD / Each
COMPARE
Chrome-plated brass 20 Yes
$224.70USD / Each
COMPARE
Chrome-plated brass 100 Yes
$224.70USD / Each
COMPARE
Chrome-plated brass 500 Yes
$224.70USD / Each
COMPARE
316 SS 20 Yes
$331.80USD / Each
COMPARE
316 SS 500 Yes
$331.80USD / Each
COMPARE
Aluminum 1200 No
$196.60USD / Each
COMPARE
316 SS 1200 No
$308.40USD / Each
COMPARE
Aluminum 1200 Yes
$211.70USD / Each
Showing 1 - 20 of 38