Masterflex® Panel-Mounted Direct-Reading Variable-Area Flowmeters with Valve for Air and Water, 65-mm Scale
No more eye strain - front shield magnifies scale 16 percent for easier readings
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
$135.40 - $282.90USD / Each
56 variations of this product are available.
See All Product Options Below
56 PRODUCT OPTIONS
Compareitem specifications
Fluid Type Valve Description
Availability Pricing
COMPARE
For Water No 65-mm Direct Reading Flowmeter w/ Valve, SS, 750 mL/min H2O
$264.60USD / Each
COMPARE
For Air Yes 65-mm Flowmeter, Aluminum/Carboloy Float; For Air, 250 mL/min
$136.40USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Aluminum/Glass Float; For Air, 1 LPM
$136.40USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Aluminum/Glass Float; For Air, 5 LPM 32013-13
$136.40USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Aluminum/Glass Float; For Air, 7 mL/min
$136.40USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Aluminum/Glass Float; For Air, 6 scfh
$136.40USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Aluminum/Ss Float; For Air, 10 scfh
$136.40USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter For Air; Aluminum With Valve, 50 scfh
$136.40USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Aluminum/Ss Float; For Air, 90 scfh
$144.30USD / Each
COMPARE
For Air Yes 65-mm Flowmeter, Brass/Ss Float; For Air, With Valve; 50 mL/min
$179.50USD / Each
COMPARE
For Air Yes 65-mm Flowmeter, Brass/Ss Float; For Air, With Valve; 10 scfh
$179.50USD / Each
COMPARE
For Water Yes 65-mm Direct Reading Flowmeter, Brass/Ss Float; For Water, 6 mL/min
$179.50USD / Each
COMPARE
For Air Yes 65-mm Flowmeter, Ss/Ss Float; For Air, With Valve; 50 mL/min
$264.60USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Ss/Carboloy Float;For Air, 250 mL/min
$264.60USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Ss/Carboloy Float;For Air, 500 mL/min 32015-07
$264.60USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Ss/Glass Float;For Air, With Valve; 5 LPM
$264.60USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Ss/Ss Float;For Air, With Valve; 10 LPM
$264.60USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Ss/Ss Float;For Air, With Valve; 10 scfh
$264.60USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter, Ss/Ss Float;For Air, With Valve; 25 scfh
$264.60USD / Each
COMPARE
For Air Yes 65-mm Direct Reading Flowmeter For Air; 316 Ss With Valve,50 scfh
$264.60USD / Each
Showing 1 - 20 of 56