Masterflex® Peristaltic Injector Pumps
A cost-effective pump for pressure injection and chemical feed applications
  • Flow rates from 6 to 250 mL/min
  • Valox® housing and tamper-resistant control cover
  • High-pressure pumps are self priming
  • Digital keypad with LED confirms pump settings and operation
  • Built-in indicator alerts you when it is time to change tubing
  • Variable-sp
With no internal check valves, these peristaltic pumps are not prone to clogging or fouling—ideal in applications with tough-to-handle solutions. Pump tubing assemblies last up to 1000 hours and take only minutes to change.
The variable-speed models ens
$716.00 - $1,241.50USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Power (VAC) Description
Availability Pricing
COMPARE
6 115 Peristaltic Injector Pump, Fixed Speed 2.3 GPD,115 VAC
$726.25USD / Each
COMPARE
25 115 Peristaltic Injector Pump, Fixed Speed 9.5 GPD, 115 VAC
$716.00USD / Each
COMPARE
63 115 Peristaltic Injector Pump, Fixed Speed 24.0 GPD, 115 VAC
$716.00USD / Each
COMPARE
250 115 Peristaltic Injector Pump, Fixed Speed 95.1 GPD, 115 VAC
$716.00USD / Each
COMPARE
200 220 Peristaltic Injector Pump, Fixed Speed 76.1 GPD, 220 VAC
$716.00USD / Each
COMPARE
6 115 Peristaltic Injector Pump, Variable Speed 2.3 GPD, 115 VAC
$1,241.50USD / Each
COMPARE
25 115 Peristaltic Injector Pump, Variable Speed 9.5 GPD, 115 VAC
$1,231.00USD / Each
COMPARE
63 115 Peristaltic Injector Pump, Variable Speed 24.0 GPD, 115 VAC
$1,231.00USD / Each
COMPARE
50 220 Peristaltic Injector Pump, Variable Speed 19.2 GPD, 220 VAC
$1,231.00USD / Each
COMPARE
138 115 Peristaltic Injector Pump, Variable Speed 52.5 GPD,115 VAC
$1,231.00USD / Each
COMPARE
250 115 Peristaltic Injector Pump, Variable Speed 95.1 GPD, 115 VAC
$1,231.00USD / Each
Showing 1- 11 of 11