Masterflex® Piston Pump Systems
These popular drive and head combinations simplify ordering!
  • Flow rates from 30 to 432 mL/min
  • Prebuilt systems with the most popular head and drive combinations
  • High repeatability and low maintenance
Series Q pump heads have no valves for a drift-free operation—providing millions of maintenance-free cycles. There is only one moving part, resulting in accuracy better than 1%.
Flow rate adjustment could not be easier: simply move the pump head position
$2,574.00 - $2,750.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Power (VAC)
Availability Pricing
COMPARE
30 100 115 to 260
$2,737.50USD / Each
COMPARE
192 100 190 to 260
$2,750.50USD / Each
COMPARE
432 50 190 to 260
$2,574.00USD / Each