This product is now discontinued, View Related Products
Masterflex® Piston Pumps, Fixed Speed, for Series Q Pump Heads
Drives are suitable for use with Series Q pump heads
  • Drive supports rpm from 2 to 60
  • Interchangeable with all Series Q pump heads
  • Offer better than ±1% accuracy.
Masterflex® L/S® fixed-Speed AC drives include the Masterflex adapter kit, part number 07104-45, needed to connect Series Q piston pump heads. The drives are interchangeable with all Series Q pump heads. Set the flow anywhere from 0 to 100% of the maximum (set speed) — drives offer better than ±1% accuracy. Pump heads sold separately.
Call For Price
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Motor RPM (rpm) Power (VAC) Description
Availability Pricing
COMPARE
2 115 Piston Pump Fixed Speed Drive for Q Pump Heads, 2 Revolutions per minute, 115 VAC
Call For Price
COMPARE
6 115 Piston Pump Fixed Speed Drive for Q Pump Heads, 6 Revolutions per minute, 115 VAC
Call For Price
COMPARE
12 115 Piston Pump Fixed Speed Drive for Q Pump Heads, 12 Revolutions per minute, 115 VAC
Call For Price
COMPARE
20 115 Piston Pump Fixed Speed Drive for Q Pump Heads, 20 Revolutions per minute, 115 VAC
Call For Price
COMPARE
60 115 Piston Pump Fixed Speed Drive for Q Pump Heads, 60 Revolutions per minute, 115 VAC
Call For Price
COMPARE
2 230 Piston Pump Fixed Speed Drive for Q Pump Heads, 2 Revolutions per minute, 230 VAC
Call For Price
COMPARE
5 230 Piston Pump Fixed Speed Drive for Q Pump Heads, 6 Revolutions per minute, 230 VAC
Call For Price
COMPARE
10 230 Piston Pump Fixed Speed Drive for Q Pump Heads, 12 Revolutions per minute, 230 VAC
Call For Price
COMPARE
17 230 Piston Pump Fixed Speed Drive for Q Pump Heads, 20 Revolutions per minute, 230 VAC
Call For Price
COMPARE
50 230 Piston Pump Fixed Speed Drive for Q Pump Heads, 60 Revolutions per minute, 230 VAC
Call For Price
Showing 1- 10 of 10