Masterflex® Plastic Tubing Clamp Ties
Special curved clamp tie head ensures tight seal
  • Ideal for general-purpose, single-use applications
Perfect for flexible fuel line, water line, and vacuum line applications. Ties measure 8" L x 0.18" W; maximum diameter of 1-3/4".
$44.25 - $44.50USD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Minimum ID (in) Description
Availability Pricing
COMPARE
1.472 Plastic Clamp Ties, White with Curved Head, 100/Pk
$44.25USD / Pkg of 100
COMPARE
0.256 Plastic Clamp Ties, Black UV Resistant with Curved Head, 100/Pk
$44.50USD / Pkg of 100