Masterflex® Pressure Booster Pump Systems
Pump system automatically adjusts speed to maintain set pressure
  • Maximum flow range of 25 to 100 GPM│NEMA 1 or NEMA 4 variable frequency drive controller with corresponding main disconnect
  • Three-phase input line voltage (single-phase systems are available)
Pump supply booster systems feature a stainless steel centrifugal pump. System is controlled by the variable frequency drive (VFD) which features an LCD and adjustable pressure range; choose the VFD with either a NEMA 1 or NEMA 4 rated enclosure and main disconnect.
System has a small footprint of approximately 30" x 30" (76.2 x 76.2 cm), and has a 1.5" NPT(F) suction and discharge connection. Piping, valve, and drain bodies are made from brass to meet most needs.
Additional options are available: stainless steel piping upgrade, suction check valve (bronze), unions (bronze), low suction shutoff, and low suction shutoff with alarm. If your facility has only single phase voltage, a custom unit can be made to match. Please contact our Application Specialists for details.
$2,757.50 - $3,608.50USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Motor Enclosure Type Description
Availability Pricing
COMPARE
38 NEMA 1 Pump Booster System, 38 GPM or 63 PSI, 2 HP, NEMA 1 enclosure
$2,757.50USD / Each
COMPARE
93 NEMA 1 Pump Booster System, 93 GPM or 44 PSI, 3 HP, NEMA 1 enclosure
$3,004.00USD / Each
COMPARE
100 NEMA 1 Pump Booster System, 100 GPM or 56 PSI, 3 HP, NEMA 1 enclosure
$3,188.50USD / Each
COMPARE
38 NEMA 4 Pump Booster System, 38 GPM or 63 PSI, 2 HP, NEMA 4 enclosure
$3,242.50USD / Each
COMPARE
100 NEMA 4 Pump Booster System, 100 GPM or 56 PSI, 3 HP, NEMA 4 enclosure
$3,608.50USD / Each