Masterflex® Printed Agglutination Slides
No more makeshift slides
  • Large wells clearly show reaction results
  • Hydrophobic epoxy ink withstands frequent washing and resists chemicals
  • Choose numbered/unnumbered—Unnumbered slides allow you to customize the labeling
Use these printed slides to perform latex and other agglutination tests for quick microbe identification. Choose between eight or twelve wells, square or circular, and numbered or unnumbered to suit your application.
$174.00 - $189.00USD / Pkg of 144
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Number of Wells Well Shape
Availability Pricing
COMPARE
8 Square
$174.00USD / Pkg of 144
COMPARE
8 Round
$187.00USD / Pkg of 144
COMPARE
12 Round
$189.00USD / Pkg of 144
COMPARE
12 Round
$189.00USD / Pkg of 144