Masterflex® Pump Handheld Remote Controllers
Easily control your Masterflex® L/S® pumps while filling and dispensing
  • Suitable for use with Masterflex® L/S® pumps
  • Long 10ft cable makes remote control simple
These ergonomic handheld controllers are designed to control basic pump drive operations including start/stop, direction, and prime — depending on available drive functions. Route tubing through the handle for convenience in filling and dispensing applications. These handheld controllers accept Masterflex tubing sizes L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18, L/S 15, and L/S 24 (up to 7/16" nominal OD).
$370.00 - $378.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Handheld Remote Controller; DB25(M) Connector; 6 ft (1.8 m) Cable
$378.00USD / Each
COMPARE
Handheld Remote Controller; DB9(M) Connector; 6 ft (1.8 m) Cable
$378.00USD / Each
COMPARE
Handheld Remote Controller; Stripped Wire Ends; 6 ft (1.8 m) Cable
$370.00USD / Each