Masterflex® Reducer Fittings, Hose Barb, Straight (Polypropylene)
Best for low-pressure applications
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range 9 to 180°F (-13 to 66°C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
$15.30 - $189.00USD / Pkg of 10
52 variations of this product are available.
See All Product Options Below
52 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
3/8 3/8 Polypropylene
$36.50USD / Pkg of 10
COMPARE
5/8 5/8 Polypropylene
$21.75USD / Pkg of 10
COMPARE
1/2 1/2 Polypropylene
$27.50USD / Pkg of 10
COMPARE
5/16 1/4 Polypropylene
$24.25USD / Pkg of 10
COMPARE
3/8 1/4 Polypropylene
$27.50USD / Pkg of 10
COMPARE
3/8 5/16 Polypropylene
$20.00USD / Pkg of 10
COMPARE
1/2 5/16 Polypropylene
$20.50USD / Pkg of 10
COMPARE
1/2 3/8 Polypropylene
$27.00USD / Pkg of 10
COMPARE
5/8 3/8 Polypropylene
$21.25USD / Pkg of 10
COMPARE
5/8 1/2 Polypropylene
$21.75USD / Pkg of 10
COMPARE
1/4 3/16 Polypropylene
$141.00USD / Pkg of 100
COMPARE
1/2 3/16 Polypropylene
$189.00USD / Pkg of 100
COMPARE
5/8 1/4 Polypropylene
$124.00USD / Pkg of 100
COMPARE
5/8 1/2 Polypropylene
$120.00USD / Pkg of 100
COMPARE
3/4 3/8 Polypropylene
$98.00USD / Pkg of 50
COMPARE
1 1/2 Polypropylene
$125.00USD / Pkg of 50
COMPARE
1 5/8 Polypropylene
$125.00USD / Pkg of 50
COMPARE
1/4 1/8 Polypropylene
$22.50USD / Pkg of 10
COMPARE
3/32 1/16 Polypropylene
$24.50USD / Pkg of 25
COMPARE
1/8 3/32 Polypropylene
$152.00USD / Pkg of 250
Showing 1 - 20 of 52