Masterflex® Reducer Fittings, Hose Barb, Straight (PVDF)
Best for low-pressure applications
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Temperture range -80 to 265°F (-62 to 129°C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
$12.92 - $85.20USD / Pkg of 10
40 variations of this product are available.
See All Product Options Below
40 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
COMPARE
3/4 1/2 PVDF
$67.50USD / Pkg of 10
COMPARE
3/8 5/16 PVDF
$55.50USD / Pkg of 10
COMPARE
1/2 3/8 PVDF
$51.00USD / Pkg of 10
COMPARE
5/8 1/4 PVDF
$63.50USD / Pkg of 10
COMPARE
3/4 3/8 PVDF
$38.50USD / Pkg of 5
COMPARE
3/4 1/2 PVDF
$40.00USD / Pkg of 5
COMPARE
3/32 1/16 PVDF
$12.92USD / Pkg of 10
COMPARE
1/8 3/32 PVDF
$12.92USD / Pkg of 10
COMPARE
1/4 5/32 PVDF
$36.80USD / Pkg of 10
COMPARE
5/16 1/4 PVDF
$35.16USD / Pkg of 10
COMPARE
3/8 5/16 PVDF
$37.84USD / Pkg of 10
COMPARE
1/2 5/16 PVDF
$44.96USD / Pkg of 10
COMPARE
1/2 3/8 PVDF
$45.84USD / Pkg of 10
COMPARE
5/8 3/8 PVDF
$50.32USD / Pkg of 10
COMPARE
5/8 1/2 PVDF
$51.20USD / Pkg of 10
COMPARE
3/4 3/8 PVDF
$59.60USD / Pkg of 10
COMPARE
3/4 1/2 PVDF
$62.00USD / Pkg of 10
COMPARE
1 3/4 PVDF
$85.20USD / Pkg of 10
COMPARE
1 1/2 PVDF
$77.60USD / Pkg of 10
COMPARE
3/32 1/16 PVDF
$27.50USD / Pkg of 10
Showing 1 - 20 of 40