Masterflex® Sanitary Adapters
Sanitary fitting with built-in gasket makes sanitary installation easier than ever
  • Sanitary gasket combined with a sanitary fitting adds another layer of safety to installation
  • Prevent contamination with zero user contact with the gasket to ensure sterility
  • Eliminate the risk of misalignment with the limited slip design
Allowing for easy assembly and disassembly of sanitary set-ups, these adapters are suitable for use connecting a Tri-Clamp® fitting to flexible tubing with the hose barb adapter. These sanitary fittings feature a built-in elastomeric gasket, providing an all-in-one singular Tri-Clamp fitting. This unique design prevents contamination by requiring zero contact between the user and the gasket. The built-in gasket also allows for a greater seal with no risk of misalignment.
Made from animal-derived ingredient-free polypropylene with a sealable/weldable TPE gasket, these adapters offer good chemical resistance and may be sterilized by gamma irradiation, autoclave, or ethylene oxide (EtO). TPE is designed specifically for cell media transfer and aseptic applications. This material offers excellent barrier properties and low protein binding.
$131.50USD / Bag of 10
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Sanitary Clamp Size (Port 1) (in) Description
Availability Pricing
COMPARE
3/4" mini clamp x 1/8" hose barb 3/4" mini Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 3/4" mini clamp x 1/8" hose barb; 10/pk
$131.50USD / Bag of 10
COMPARE
3/4" mini clamp x 3/16" hose barb 3/4" mini Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 3/4" mini clamp x 3/16" hose barb; 10/pk
$131.50USD / Bag of 10
COMPARE
3/4" mini clamp x 1/4" hose barb 3/4" mini Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 3/4" mini clamp x 1/4" hose barb; 10/pk
$131.50USD / Bag of 10
COMPARE
3/4" mini clamp x 3/8" hose barb 3/4" mini Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 3/4" mini clamp x 3/8" hose barb; 10/pk
$131.50USD / Bag of 10
COMPARE
3/4" mini clamp x 1/2" hose barb 3/4" mini Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 3/4" mini clamp x 1/2" hose barb; 10/pk
$131.50USD / Bag of 10
COMPARE
3/4" mini clamp x 3/4" hose barb 3/4" mini Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 3/4" mini clamp x 3/4" hose barb; 10/pk
$131.50USD / Bag of 10
COMPARE
1-1/2 clamp x 1/4" hose barb 1-1/2" Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 1-1/2 clamp x 1/4" hose barb; 10/pk
$131.50USD / Bag of 10
COMPARE
1-1/2 clamp x 3/8" hose barb 1-1/2" Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 1-1/2 clamp x 3/8" hose barb; 10/pk
$131.50USD / Bag of 10
COMPARE
1-1/2 clamp x 1/2" hose barb 1-1/2" Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 1-1/2 clamp x 1/2" hose barb; 10/pk
$131.50USD / Bag of 10
COMPARE
1-1/2 clamp x 3/4" hose barb 1-1/2" Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 1-1/2 clamp x 3/4" hose barb; 10/pk
$131.50USD / Bag of 10
COMPARE
1-1/2 clamp x 1" hose barb 1-1/2" Sanitary Barbed Adapter, Polypropylene with TPE Fused Gasket, 1-1/2 clamp x 1" hose barb; 10/pk
$131.50USD / Bag of 10
Showing 1- 11 of 11