Masterflex® Sanitary Fitting Clamps, PVDF
Maintain sanitary conditions in your processes
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These sanitary clamps are compatible with Tri-Clamp® fittings. The PVDF material has excellent chemical resistance with low leachable characteristics.
$32.50 - $93.50USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Description
Availability Pricing
COMPARE
PVDF 1/2 Push/pull clamp; PVDF for sanitary fittings
$32.50USD / Each
COMPARE
PVDF 3/4 PVDF Sanitary Clamp, 3/4" Mini —40 to 280° F
$93.50USD / Each
COMPARE
PVDF 1-1/2 PVDF Sanitary Clamp, 1" and 1 1/2" —40 to 280° F
$67.50USD / Each
COMPARE
PVDF 2 PVDF Sanitary Clamp, 2" —40 to 280° F
$69.50USD / Each
COMPARE
PVDF 2-1/2 PVDF Sanitary Clamp, 2 1/2" —40 to 280° F
$87.00USD / Each