Masterflex® Sanitary Fitting End Caps
Close off your sanitary fluid line while creating a view port
  • Create a viewing port into your sanitary application with transparent end caps
  • Constructed of polysulfone for enhanced strength and durability
  • Fully compatible with other Tri-Clamp® connections
These end caps play an important role in the sanitation process, allowing an open end to be sanitary sealed preventing additional debris from accumulating while creating a view port for sample viewing. Choose from a wide range of end cap sizes, ranging from 1½” to 8”.
$174.00 - $437.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Connection Type Sanitary Clamp Size (Port 1) (in)
Availability Pricing
COMPARE
Polysulfone Sanitary 1.5
$174.00USD / Each
COMPARE
Polysulfone Sanitary 2
$174.00USD / Each
COMPARE
Polysulfone Sanitary 2.5
$181.00USD / Each
COMPARE
Polysulfone Sanitary 3
$209.00USD / Each
COMPARE
Polysulfone Sanitary 4
$241.00USD / Each
COMPARE
Polysulfone Sanitary 6
$419.00USD / Each
COMPARE
Polysulfone Sanitary 8
$437.00USD / Each
Showing 1- 7 of 7