This product is now discontinued, View Related Products
Masterflex Self-Contained Composite Sampler
  • Fixed-speed motor and built-in timer for unattended operation
This portable pump samples water, slurries, and liquid wastes at depths to 25 ft (7.6 m). Before each cycle, the system reverses flow purging the tubing and unclogging debris from the inlet. Samples only contact the tubing and stainless steel fittings.
Select 15- or 30-minute sampling intervals, or continuous operation. Solid-state electronic timer ensures precise cycling; test button allows you to obtain a single test sample. Sample volume obtainable at any setting is inversely proportional to sampling depth and tubing length.
Front panel indicator alerts you when the built-in batteries need recharging. Recharging takes 10 to12 hours. Use the optional AC adapter 07578-11 for 115 VAC operation (230 VAC operation requires adapter 01578-02 sold separately).
Sampler includes MASTERFLEX(R) drive, pump head, MASTERFLEX(R) L/S(R) 15 silicone tubing for the pump head, batteries, and a 2-1/2-gallon polyethylene container - all in a sturdy case that measures 16-5/8"W x 25-1/4"H x 9-5/8"D (42.2 cm W x 64.1 cm H x 24.5 cm D). Sampling line tubing is not included, use your own or order separately. Replacement pump head tubing can also be ordered separately.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem Availability Pricing