Masterflex® Single-Syringe Infusion Pumps
Ideal for low-flow, low-pressure applications where precision is needed
  • Flow rate of 0.0033 to 8.33 mL/min per syringe
  • Backlit alphanumeric LCD guides you through setup and operating procedures
  • Nonvolatile memory retains settings even after your pump is turned off
  • Accurate to +/-0.5%
This features microprocessor motor control and precision gearing. Microprocessor automatically selects display units based on syringe used. Obtain precise flow rates down to 0.2 µL/hr. A wide dynamic range (6000:1) provides excellent flexibility. All comm
$1,635.00 - $1,765.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate Per Syringe (mL/min) Min Flow Rate Per Syringe (microL/min) Power (VAC)
Availability Pricing
COMPARE
8.33 0.0033 115
$1,635.00USD / Each
COMPARE
8.33 0.0033 230
$1,765.50USD / Each