Masterflex® Single-Use Forceps
No more cross-contamination
  • Protect yourself and your samples with disposable forceps
  • Autoclavable plastic or stainless steel construction
These single-use forceps are bulk-packed for convenience and affordability. Slide forceps feature an extra-wide grip for greater stability and security when staining cells and tissue. Fine-tipped forceps are ideally suited for work with smaller specimens. Stainless steel forceps have grooved finger grips and ridged tips, best for larger, soft or wet specimens.
$94.50 - $120.00USD / Pkg of 25
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Overall Length (in) Sterile
Availability Pricing
COMPARE
Plastic 4 1/2 No
$120.00USD / Pkg of 50
COMPARE
Stainless steel 4 1/2 Yes
$94.50USD / Pkg of 25
COMPARE
Plastic 4 No
$102.00USD / Pkg of 50
Showing 1- 3 of 3
Need Help with this product?
Call Us At1-847-549-7600
Live chat