Masterflex® Single-Use I/P® Gamma-Irradiated Precision Pump Tubing
Reduce the risk of microbial contamination
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Gamma sensitive labeling confirms treatment
Masterflex® gamma-irradiated pump tubing protects your sensitive processes and products from accidental contamination while saving time! Gamma-irradiated tubing helps you avoid costly delays that can be associated with outsourced gamma irradiation services. Tubing is double bagged prior to undergoing the gamma irradiation sterilization process, and all sealed tubing is gamma irradiated at 32.5 kGy to offer optimal microbial control in a critical fluid pathway. Tubing boxes include gamma radiation-sensitive labels to confirm treatment. Ideal for pharmaceutical and biopharmaceutical processing, sensitive research, single-use systems, and more.
Available formulations include:
Platinum-Cured Silicone
PharMed BPT
C-Flex (001)
$248.00 - $619.00USD / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
I/P Masterflex tubing size Tubing formulation Description
Availability Pricing
COMPARE
73 Platinum-Cured Silicone I/P® Precision Pump Tubing, Gamma-Irradiated Platinum-Cured Silicone, I/P 73; 25 ft
$400.00USD / Pkg of 1
COMPARE
82 Platinum-Cured Silicone I/P® Precision Pump Tubing, Gamma-Irradiated Platinum-Cured Silicone, I/P 82; 25 ft
$619.00USD / Pkg of 1
COMPARE
73 PharMed® BPT I/P® Precision Pump Tubing, Gamma-Irradiated PharMed® BPT, I/P 73; 25 ft
$461.00USD / Pkg of 1
COMPARE
82 PharMed® BPT I/P® Precision Pump Tubing, Gamma-Irradiated PharMed® BPT, I/P 82; 25 ft
$535.00USD / Pkg of 1
COMPARE
73 C-Flex® I/P® Precision Pump Tubing, Gamma-Irradiated C-Flex®, I/P 73; 25 ft
$248.00USD / Pkg of 1
COMPARE
82 C-Flex® I/P® Precision Pump Tubing, Gamma-Irradiated C-Flex®, I/P 82; 25 ft
$284.00USD / Pkg of 1