Masterflex® Single-Use Transfer Tubing, Gamma-Irradiated C-Flex® Clear

Reduce the risk of microbial contamination
  • Ideal for general -purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Gamma sensitive labeling confirms treatment
  • Ultra smooth inner surface results in fewer extractables
Masterflex® gamma-irradiated transfer tubing protects your sensitive processes and products from accidental contamination while saving time! Gamma-irradiated tubing helps you avoid costly delays that can be associated with outsourced gamma irradiation services. Tubing is double bagged prior to undergoing the gamma irradiation sterilization process, and all sealed tubing is gamma irradiated at 32.5 kGy to offer optimal microbial control in a critical fluid pathway. Tubing boxes include gamma radiation-sensitive labels to confirm treatment. Ideal for pharmaceutical and biopharmaceutical processing, sensitive research, single-use systems, and more.

Masterflex® C-Flex® tubing offers enhances biocompatibility with longer pumping life. This tubing is nonpyrogenic, noncytotoxic, and nonhemolytic — protecting the purity of your fluid. Heat sealable and weldable, C-Flex tubing is ideal for biopharmaceutical processing and storage and botanical production.
$101.00 - $369.00USD / Pkg of 1

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hose Barb Size (in) Tubing OD (mm) Description
Availability Pricing
COMPARE
1/8 6.35 Single-Use Transfer Tubing, Gamma-Irradiated C-Flex® Clear, 1/8" ID x 1/4" OD; 25 ft
$101.00USD / Pkg of 1
COMPARE
1/4 11.1125 Single-Use Transfer Tubing, Gamma-Irradiated C-Flex® Clear, 1/4" ID x 7/16" OD; 25 ft
$179.00USD / Pkg of 1
COMPARE
3/8 15.875 Single-Use Transfer Tubing, Gamma-Irradiated C-Flex® Clear, 3/8" ID x 5/8" OD; 25 ft
$265.00USD / Pkg of 1
COMPARE
1/2 19.05 Single-Use Transfer Tubing, Gamma-Irradiated C-Flex® Clear, 1/2" ID x 3/4" OD; 25 ft
$369.00USD / Pkg of 1
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat