Masterflex® Solenoid-Operated Pinch Valves
No contamination of valve
  • Control flow in your tubing without direct contact between the fluid and the valve
  • Fast response time
  • Low-power consumption and high-cycle life
  • Excellent for sanitary and pharmaceutical applications
Choose from two-way NC, two-way NO, or three-way pinch valves. These are ideal where contamination is an issue, as the fluid only comes in contact with the tubing, not the valve. Tubing replacement is quick and easy, making these suitable for pharmaceutical or sanitary applications. Mounting clip or mounting flange available separately for increased versatility.
$122.00 - $223.00USD / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
Compareitem specifications
Flow Pattern Pinch Valve Tube Size Power (VDC)
Availability Pricing
COMPARE
Two-Way, Normally Closed 1/32" ID x 3/32" OD 12
$122.00USD / Each
COMPARE
Two-Way, Normally Closed 1/16" ID x 1/8" OD 12
$122.00USD / Each
COMPARE
Two-Way, Normally Closed 0.0625 in ID x 0.1875 in OD 12
$157.00USD / Each
COMPARE
Two-Way, Normally Closed 1/8" ID x 1/4" OD 12
$193.00USD / Each
COMPARE
Two-Way, Normally Closed 1/16" ID x 1/8" OD 24
$129.00USD / Each
COMPARE
Two-Way, Normally Closed 1/8" ID x 1/4" OD 24
$193.00USD / Each
COMPARE
Two-Way, Normally Open 1/32" ID x 3/32" OD 12
$137.00USD / Each
COMPARE
Two-Way, Normally Open 1/16" ID x 1/8" OD 12
$137.00USD / Each
COMPARE
Two-Way, Normally Open 0.0625 in ID x 0.1875 in OD 12
$154.00USD / Each
COMPARE
Two-Way, Normally Closed 1/8" ID x 1/4" OD 12
$216.00USD / Each
COMPARE
Two-Way, Normally Open 1/32" ID x 3/32" OD 24
$136.00USD / Each
COMPARE
Two-Way, Normally Open 1/16" ID x 1/8" OD 24
$135.00USD / Each
COMPARE
0.0625 in ID x 0.1875 in OD 24
$154.00USD / Each
COMPARE
Two-Way, Normally Closed 1/8" ID x 1/4" OD 24
$214.00USD / Each
COMPARE
Three-Way, One Normally Open, One Normally Closed 1/32" ID x 3/32" OD 12
$158.00USD / Each
COMPARE
0.0625 in ID x 0.1875 in OD 12
$182.00USD / Each
COMPARE
Three-Way, One Normally Open, One Normally Closed 1/32" ID x 3/32" OD 24
$159.00USD / Each
COMPARE
Three-Way, One Normally Open, One Normally Closed 1/16" ID x 1/8" OD 24
$159.00USD / Each
COMPARE
0.0625 in ID x 0.1875 in OD 24
$182.00USD / Each
COMPARE
Three-Way, One Normally Open, One Normally Closed 1/8" ID x 1/4" OD 24
$219.00USD / Each
Showing 1 - 20 of 24
Need Help with this product?
Call Us At1-847-549-7600
Live chat