Masterflex® Stopcocks for Glass and Rigid Tubing
Easily connect to rigid tubing
Stopcocks have wetted parts constructed of PFA. An O-ring made of Viton® protects the glass tube when you tighten the compression nut. Stopcocks also accept rigid tubing such as PTFE.
Available in two-way and three-way styles.
$153.00 - $277.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Tubing (mm)
Availability Pricing
COMPARE
$239.00USD / Each
COMPARE
$153.00USD / Each
COMPARE
$180.00USD / Each
COMPARE
$277.00USD / Each