Masterflex® Stopcocks with Tube Compression and NPT Connections, PTFE
Use with caustic and high-purity liquids in many applications
  • All-PTFE valve is precision machined
PTFE stopcocks function in a manner similiar to ball valves but have a tighter seal when closed, making them ideal for applications where vacuum and pressure will be used. Valve is fully open when handle is in the direction of the flow, and fully closed when it is perpendicular to the valve. At a 45° position, the valve is half open, and can thereby be used in metering applications.
$229.00 - $443.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Orifice (in) Wetted Materials
Availability Pricing
COMPARE
Compression 0.0625 PTFE
$229.00USD / Each
COMPARE
Compression 0.125 PTFE
$277.00USD / Each
COMPARE
Compression 0.25 PTFE
$284.00USD / Each
COMPARE
Compression 0.25 PTFE
$443.00USD / Each
COMPARE
Compression 0.375 PTFE
$381.00USD / Each
COMPARE
Compression 0.5 PTFE
$427.00USD / Each
COMPARE
NPT(F) 0.125 PTFE
$245.00USD / Each
COMPARE
NPT(F) 0.25 PTFE
$254.00USD / Each
COMPARE
NPT(F) 0.125 PTFE
$300.00USD / Each
COMPARE
NPT(F) 0.28 PTFE
$346.00USD / Each
COMPARE
NPT(F) 0.44 PTFE
$416.00USD / Each
Showing 1- 11 of 11