Masterflex® Submersible Sampling Pumps
12 VDC powered is ideal for those field well purging/sampling applications
  • Practical for dedicated use and disposal
These Masterflex® submersible sampling pumps are suitable for purging and sampling applications. Attach pump to 1/2” or 3/8” tubing (tubing sold separately), and lower pump directly into well. Pumps are constructed with an ABS plastic and polycarbonate body, stainless steel impeller, and nitrile seal.
$164.00 - $485.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Flow Rate (GPM)
Availability Pricing
COMPARE
6.3125 1.4
$164.00USD / Each
COMPARE
13.625 2.3
$310.00USD / Each
COMPARE
15.75 3.3
$485.00USD / Each