Masterflex® Thermal Mass Flowmeters/Flow Transmitters for Gases
Get faster performance with miniature coil design
  • All meters feature 0 to 5 VDC output signal for data logging or controlling other instruments
These low-cost flowmeters use thermal gas flow sensing technique that results in highly accurate readings and repeatability. All units are calibrated to air/nitrogen—call our Application Specialists for meters calibrated to other gases or gas mixtures. Meters feature a cable hub connector for a quick connection to a power supply and 0 to 5 VDC output cable; order power supplies/cables under accessories.
Flowmeters have low pressure drop across the sensor. The output signal can connect to a remote display, recorder, or any instrument that accepts a 0 to 5 VDC signal. Compact size makes it easy to carry around.
Wetted materials are anodized aluminum, 316 stainless steel, Viton® O-rings, and 316 stainless steel, and acetal fittings. Order rechargeable battery kit under Accessories.
$380.00 - $1,868.50USD / Each
24 variations of this product are available.
See All Product Options Below
24 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Display Type Max Flow Rate (LPM) Wetted Materials
Availability Pricing
COMPARE
None 0.1 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$433.90USD / Each
COMPARE
None 0.2 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$443.00USD / Each
COMPARE
None 0.5 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$443.00USD / Each
COMPARE
None 1 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$459.10USD / Each
COMPARE
None 5 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$469.60USD / Each
COMPARE
None 10 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$523.25USD / Each
COMPARE
3.5-digit LCD 0.02 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$709.75USD / Each
COMPARE
3.5-digit LCD 0.05 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$766.75USD / Each
COMPARE
3.5-digit LCD 0.1 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$849.50USD / Each
COMPARE
3.5-digit LCD 0.2 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$857.50USD / Each
COMPARE
3.5-digit LCD 0.5 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$834.00USD / Each
COMPARE
3.5-digit LCD 1 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$836.50USD / Each
COMPARE
3.5-digit LCD 2 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$849.50USD / Each
COMPARE
3.5-digit LCD 5 Acetal, Viton, Anodized Aluminum, 316 Stainless Steel
$829.50USD / Each
COMPARE
3.5-digit LCD 10 Viton, Anodized Aluminum, 316 Stainless Steel
$911.50USD / Each
COMPARE
None 20 Viton, Anodized Aluminum, 316 Stainless Steel
$823.75USD / Each
COMPARE
None 50 Viton, Anodized Aluminum, 316 Stainless Steel
$1,011.00USD / Each
COMPARE
3.5-digit LCD 20 Viton, Anodized Aluminum, 316 Stainless Steel
$1,228.00USD / Each
COMPARE
3.5-digit LCD 100 Viton, Anodized Aluminum, 316 Stainless Steel
$1,463.50USD / Each
COMPARE
3.5-digit LCD 500 Viton, Anodized Aluminum, 316 Stainless Steel
$1,868.50USD / Each
Showing 1 - 20 of 24