Masterflex® Touch-Screen, Continuous-Cycle, Four-Syringe Pumps
Continuously cycle up to four syringes in a push-pull action
  • Flow range from 5 pL/min to 215.8 mL/min
  • Intuitive touch screen allows you to quickly create and recall configurations
  • You can daisy chain up to 99 pumps together using the built in RS-485
  • Accuracy of ±0.35% and reproducibility of ±0.05%
Hold up to four syringes to cycle continuously back and forth in a push-pull action. As two syringes are infusing, two syringes are simultaneously withdrawing at the same rate. At the end of the set volume the direction is automatically reversed and the n
$4,655.00 - $4,663.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate Per Syringe (mL/min) Min Flow Rate Per Syringe (microL/min) Number of Syringes
Availability Pricing
COMPARE
215.8 4
$4,663.00USD / Each
COMPARE
215.8 4
$4,655.00USD / Each